Assay Details
| Gene Symbol: Zfp955b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142957.1 | NP_001136429.1 | 4 | 2244 | 190 | |
| GenBank mRNA | AK161843.1 | - | 4 | 2245 | 190 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P681705 | AK135959.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P681705 | NM_001142957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P681705 | AK020746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P681705 | AK135959.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P681705 | NM_001142957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P681705 | AK020746.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177991 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150460 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P681705 | AK135959.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P681705 | NM_001142957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P681705 | AK020746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P681705 | AK135959.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P681705 | NM_001142957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P681705 | AK020746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681705 | AK135959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681705 | NM_001142957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681705 | AK020746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681705 | AK135959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681705 | NM_001142957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681705 | AK020746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P470462 | AK083028.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P470462 | AK050454.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148662 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443944 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453198_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429560_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443940 | - | - |
| Mouse Genome 430 2.0 Array | 1453198_at | - | - |
| Mouse Genome 430 2.0 Array | 1429560_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453198_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429560_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100043468_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100043468_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700005256.mm.1 | NM_001142957.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005256.mm.1 | NM_001142957.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005256.mm.1 | TC1700000580.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005257.mm.1 | NM_001142957.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005257.mm.1 | NM_001142957.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005257.mm.1 | TC1700000580.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429560_PM_at | - | - |
| Mouse Expression Set 430 | 1453198_PM_at | - | - |
| Mouse Expression Set 430 | 1429560_at | - | - |
| Mouse Expression Set 430 | 1453198_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453198_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429560_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429560_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453198_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453198_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429560_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429560_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453198_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 490554 | - | - |

Zfp955b
Genomic Map