Assay Details
| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053152.2 | NP_444382.1 | 1 | 50 | 98 | |
| GenBank mRNA | AF288381.1 | - | 1 | 50 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF288381.1, NM_053152.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170350 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852892 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56285 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122043 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174017 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112911 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108063 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170349 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170350 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446311 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446275 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446339 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069860 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112911 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446337 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069325 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069737 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069147 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069621 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069429 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446299 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446293 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069777 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069244 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069208 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174017 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446317 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069506 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069642 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069589 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069086 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069108 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123852 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069636 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446282 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446341 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069619 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069884 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108063 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446308 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446306 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446344 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069196 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069247 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069134 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069473 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446350 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069385 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446347 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01446287 | - | - |
| Whole Mouse Genome G4122A | A_51_P132079 | NM_053152.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132079 | AF288381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132079 | AF419250.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069385 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069589 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01446313 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069671 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122043 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174017 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069619 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123852 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069473 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01446299 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01446347 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069147 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01446337 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069621 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069196 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069737 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01446317 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01446275 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069325 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069086 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06069860 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108063 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112911 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05654496 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05480443 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05654499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05839148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05061571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05381629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05654499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05061573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05061577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548491 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548457 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548455 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608659 | - | - |
| Mouse Genome 430 2.0 Array | 1452645_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452645_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025519.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025516.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025516.mm.1 | NM_016659.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025516.mm.1 | NM_016659.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025517.mm.1 | TC0600003607.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025517.mm.1 | NM_016659.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025517.mm.1 | NM_016659.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025436.mm.1 | TC0600003607.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452645_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452645_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452645_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452645_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 476267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 100130427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370746 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664429 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233141 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664429 | - | - |
| mouse-6 v1.1 (Illumina) | scl0093969.1_46-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_21361217-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370746 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100130427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0093969.1_46-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100130427 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2664429 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233141 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664429 | - | - |

Klra22
Genomic Map