Assay Details
| Gene Symbol: Cldn34b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081671.1 | NP_001075140.1 | 3 | 856 | 128 | |
| GenBank mRNA | BC100353.1 | - | 3 | 856 | 30933439 | 128 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093189 | NM_001081671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093189 | AK006680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093189 | BC100353.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456626 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750828 | AK034177.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750828 | BC034128.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750828 | BC057086.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750828 | NM_207222.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63443 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093189 | NM_001081671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093189 | AK006680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093189 | BC100353.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09465623 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09465612 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03780353 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09465600 | - | - |
| Whole Mouse Genome G4122A | A_51_P486320 | NM_001081671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486320 | AK006680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486320 | BC100353.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118037 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09465623 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09465600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32758509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15051393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14878807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14974430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14870642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14719440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14941472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15051402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32758583 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607461 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432326_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607459 | - | - |
| Mouse Genome 430 2.0 Array | 1432326_at | - | - |
| Mouse Genome 430 2.0 Array | 1432326_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00020237.mm.1 | NM_001081671.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020237.mm.1 | NM_001081671.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020237.mm.1 | TC0X00003234.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00020240.mm.1 | NM_001081671.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020240.mm.1 | NM_001081671.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020240.mm.1 | TC0X00003234.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00020239.mm.1 | NM_001081671.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020239.mm.1 | NM_001081671.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020239.mm.1 | TC0X00003234.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432326_at | - | - |
| Mouse Expression Set 430 | 1432326_PM_at | - | - |
| Murine Genome U74v2 Set | 169319_r_at | - | - |
| Murine Genome U74v2 Set | 169319_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432326_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432326_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247663 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247663 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2967254 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2967254 | - | - |

Cldn34b2
Genomic Map