Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10499975 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10499971 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10499963 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0300018697.mm.1 |
NM_148949.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018697.mm.1 |
NM_148949.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018697.mm.1 |
TC0300002425.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018705.mm.1 |
NM_001163730.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018705.mm.1 |
TC0300002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018694.mm.1 |
NM_148949.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018694.mm.1 |
NM_148949.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018694.mm.1 |
TC0300002425.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018704.mm.1 |
NM_001163730.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018704.mm.1 |
TC0300002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018709.mm.1 |
NM_001163730.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018709.mm.1 |
TC0300002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018706.mm.1 |
NM_001163730.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018706.mm.1 |
TC0300002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300018695.mm.1 |
NM_148949.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018695.mm.1 |
NM_148949.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300018695.mm.1 |
TC0300002425.mm.1 |
MTA TC |
Murine Genome U74v2 Set |
132796_at |
- |
- |
Murine Genome U74v2 Set |
132796_PM_at |
- |
- |