Assay Details
| Gene Symbol: Slc39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013901.2 | NP_038929.2 | 1 - 2 | 59 | 63 | |
| GenBank mRNA | AK146175.1 | - | 1 - 2 | 51 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK146175.1,NM_013901.2 transcripts.
This assay is in the 5'UTR region of the AK146175.1,NM_013901.2 transcripts.
Set Membership: |
|

Slc39a1
Genomic Map