Assay Details
| Gene Symbol: Rfpl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177783.1 | NP_001171254.1 | 1 - 2 | 92 | 119 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY726306.1 | - | 92 | Chr. - - [-] on Build | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001177783.1 transcripts.
Set Membership: |
|

Rfpl4b
Genomic Map