Assay Details
| Gene Symbol: 9330175E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015514.2 | 1 - 2 | 274 | 94 | ||
| GenBank mRNA | AK053303.1 | - | 1 - 2 | 276 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1 |
Herpud1 |
Herp, Mifl, SUP |
-38.9kb | Anti-sense | Chr. 8 - 94386143 - 94395366 on NCBI Build GRCm38 |
| Downstream | microRNA 7072 | Mir7072 |
Gm27665, mmu-mir-7072 |
69.9kb | Anti-sense | Chr. 8 - 94505046 - 94505105 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9330175E14Rik
Genomic Map