Assay Details
| Gene Symbol: Tspy-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027507.1 | 1 - 2 | 49 | 99 | ||
| GenBank mRNA | AJ001378.1 | - | 1 - 2 | 51 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
eukaryotic translation initiation factor 2, subunit 3, structural gene Y-linked |
Eif2s3y |
Eif-2gy, Spy, Tfy |
-10.4kb | Anti-sense | Chr. Y - 1010578 - 1046811 on NCBI Build GRCm38 |
| Downstream |
ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome |
Uty |
Hydb, mKIAA4057 |
26.3kb | Sense | Chr. Y - 1085187 - 1245754 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Tspy-ps
Genomic Map