Assay Details
| Gene Symbol: Mug-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027619.1 | 16 - 17 | 2012 | 102 | ||
| GenBank mRNA | M65237.1 | - | 3 - 4 | 625 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | murinoglobulin pseudogene | Gm10319 |
EG381806, ENSMUSG00000071204 |
-80.5kb | Sense | Chr. 6 - 122136628 - 122150958 on NCBI Build GRCm38 |
| Downstream |
killer cell lectin-like receptor subfamily G, member 1 |
Klrg1 |
2F1-Ag, MAFA, MAFA-L |
37.7kb | Anti-sense | Chr. 6 - 122270596 - 122282833 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mug-ps1
Genomic Map