Assay Details
| Gene Symbol: Gm2762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037991.1 | 4 - 5 | 417 | 74 | ||
| GenBank mRNA | AK030238.1 | - | 4 - 5 | 418 | 74 | |
| BB617179.1 | - | 4 - 5 | 418 | 74 | ||
| BC100495.1 | - | 4 - 5 | 398 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
serine palmitoyltransferase, long chain base subunit 1 |
Sptlc1 |
AW552086, C77762, E030036H05, Lcb1 |
-20.5kb | Anti-sense | Chr. 13 - 53332748 - 53377417 on NCBI Build GRCm38 |
| Downstream | msh homeobox 2 | Msx2 |
BB122635, Hox-8, Hox8, Hox8.1 |
34.3kb | Anti-sense | Chr. 13 - 53466881 - 53472780 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm2762
Genomic Map