Assay Details
| Gene Symbol: Celrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038008.1 | 3 - 4 | 310 | 76 | ||
| GenBank mRNA | AK138346.1 | - | 3 - 4 | 311 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2610027F03 gene | 2610027F03Rik | -350.2kb | Sense | Chr. 1 - 120614387 - 120763567 on NCBI Build GRCm38 | |
| Downstream | insulin induced gene 2 | Insig2 |
2900053I11Rik, C730043J18Rik, Insig-2 |
186.9kb | Anti-sense | Chr. 1 - 121304353 - 121333654 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Celrr
Genomic Map