Assay Details
| Gene Symbol: Gm17751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038012.1 | 4 - 5 | 492 | 95 | ||
| GenBank mRNA | AK162144.1 | - | 4 - 5 | 492 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 4 (anion exchanger), member 3 |
Slc4a3 |
A930038D23Rik, Ae3 |
-938.1kb | Sense | Chr. 1 - 75546041 - 75559431 on NCBI Build GRCm38 |
| Downstream | microRNA 6343 | Mir6343 |
Gm27304, mmu-mir-6343 |
7.5kb | Sense | Chr. 1 - 76509560 - 76509643 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm17751
Genomic Map