Assay Details
| Gene Symbol: Zp4-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027813.1 | 5 - 6 | 702 | 102 | ||
| GenBank mRNA | DQ279480.1 | - | 5 - 6 | 702 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cholinergic receptor, muscarinic 3, cardiac |
Chrm3 |
Chrm-3, M3, M3R |
-1.2mb | Anti-sense | Chr. 13 - 9875488 - 10360803 on NCBI Build GRCm38 |
| Downstream |
ryanodine receptor 2, cardiac |
Ryr2 |
9330127I20Rik, RYR-2 |
27.9kb | Anti-sense | Chr. 13 - 11553099 - 12106945 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Zp4-ps
Genomic Map