Assay Details
| Gene Symbol: Bambi-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027919.1 | 1 | 46 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 28 (sodium-coupled nucleoside transporter), member 2 |
Slc28a2 |
2010208B10Rik, B430217P18, BB152493, Cnt2 |
-5.4kb | Anti-sense | Chr. 2 - 122426355 - 122461152 on NCBI Build GRCm38 |
| Downstream | predicted gene 14085 | Gm14085 | OTTMUSG00000015615 | 17.1kb | Anti-sense | Chr. 2 - 122484941 - 122528040 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bambi-ps1
Genomic Map