Assay Details
| Gene Symbol: AF357399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028129.1 | 1 | 44 | 84 | ||
| GenBank mRNA | AF357399.1 | - | 1 | 44 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppressor of Ty 5 | Supt5 |
AL033283, AL033321, AU019549, Spt5, Supt5h |
-12.7kb | Anti-sense | Chr. 7 - 28314891 - 28338804 on NCBI Build GRCm38 |
| Downstream |
pleckstrin homology domain containing, family G (with RhoGef domain) member 2 |
Plekhg2 |
AI194308, Clg, Cslg |
8.0kb | Anti-sense | Chr. 7 - 28359603 - 28372687 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AF357399
Genomic Map