Assay Details
| Gene Symbol: Gm5129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028426.1 | 1 | 50 | 80 | ||
| GenBank mRNA | AK076721.1 | - | 1 | 53 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 42245 |
Gm42245 | -35.3kb | Sense | Chr. 5 - 29684782 - 29699986 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 36151 |
Gm36151 | 17.7kb | Anti-sense | Chr. 5 - 29753659 - 29755678 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm5129
Genomic Map