Assay Details
| Gene Symbol: Gm15910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038023.1 | 1 - 2 | 135 | 85 | ||
| GenBank mRNA | AK135362.1 | - | 1 - 2 | 136 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methionine sulfoxide reductase B3 | Msrb3 |
D430026P16Rik, I-8-23, MsrB2 |
Anti-sense | Chr. 10 - 120781098 - 120899304 on NCBI Build GRCm38 | |
| Downstream | LEM domain containing 3 | Lemd3 |
AI316861, Man1 |
64.4kb | Anti-sense | Chr. 10 - 120923411 - 120979949 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15910
Genomic Map