Assay Details
| Gene Symbol: Gm6150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038036.1 | 2 - 3 | 385 | 101 | ||
| GenBank mRNA | AK139890.1 | - | 2 - 3 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterile alpha motif domain containing 5 | Samd5 | E130306M17Rik | -11.4kb | Anti-sense | Chr. 10 - 9627259 - 9675208 on NCBI Build GRCm38 |
| Downstream | syntaxin binding protein 5 (tomosyn) | Stxbp5 |
0710001E20Rik, 4930565N16Rik, AW742610, LGL3, mKIAA1006 |
66.7kb | Anti-sense | Chr. 10 - 9755547 - 9901227 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6150
Genomic Map