Assay Details
| Gene Symbol: Scarna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028519.2 | 1 | 152 | 98 | ||
| GenBank mRNA | AF357345.1 | - | 1 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | inositol polyphosphate-5-phosphatase D | Inpp5d |
SHIP, SHIP-1, SHIP1, SIP-145, p150Ship, s-SHIP |
-64.1kb | Sense | Chr. 1 - 87620312 - 87720510 on NCBI Build GRCm38 |
| Downstream |
S-antigen, retina and pineal gland (arrestin) |
Sag |
A930001K18Rik, Arr1, Irbp, arrestin |
18.9kb | Sense | Chr. 1 - 87803680 - 87845158 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scarna6
Genomic Map