Assay Details
| Gene Symbol: Snord11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028521.1 | 1 | 37 | 62 | ||
| GenBank mRNA | AF357320.1 | - | 1 | 37 | 62 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CB571381.1 | - | 55 | Chr. - - [-] on Build | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 70 |
Snord70 |
MBII-234, snoRNA234 |
-12.8kb | Sense | Chr. 1 - 59691958 - 59692010 on NCBI Build GRCm38 |
| Downstream |
bone morphogenetic protein receptor, type II (serine/threonine kinase) |
Bmpr2 |
2610024H22Rik, AL117858, AW546137, BB189135, BMP-2, BMPR-2, BMPR-II, BMPRII, BRK-3, Gm20272 |
59.4kb | Sense | Chr. 1 - 59764270 - 59878081 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord11
Genomic Map