Assay Details
| Gene Symbol: Snord67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028553.1 | 1 | 44 | 85 | ||
| GenBank mRNA | AF357343.1 | - | 1 | 44 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | low density lipoprotein receptor-related protein 4 | Lrp4 |
6430526J12Rik, D230026E03, Megf7, mdig |
-82.2kb | Sense | Chr. 2 - 91457470 - 91513901 on NCBI Build GRCm38 |
| Downstream | coagulation factor II | F2 |
Cf-2, Cf2, FII |
17.8kb | Anti-sense | Chr. 2 - 91613986 - 91636452 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord67
Genomic Map