Assay Details
| Gene Symbol: Gm10584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028578.1 | 1 | 2550 | 101 | ||
| GenBank mRNA | AK141912.1 | - | 1 | 2550 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 19463 |
Gm19463 | Anti-sense | Chr. 7 - 132314912 - 132316297 on NCBI Build GRCm38 | ||
| Downstream |
predicted gene, 39102 |
Gm39102 | 101.7kb | Sense | Chr. 7 - 132420030 - 132423713 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm10584
Genomic Map