Assay Details
| Gene Symbol: Gm10649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028579.1 | 1 - 2 | 68 | 75 | ||
| GenBank mRNA | AK138650.1 | - | 1 - 2 | 69 | 75 | |
| AK162930.1 | - | 1 - 2 | 69 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102640923 | LOC102640923 | Sense | Chr. 8 - 76605226 - 76899985 on NCBI Build GRCm38 | ||
| Downstream | Rho GTPase activating protein 10 | Arhgap10 |
A930033B01Rik, PSGAP-m, PSGAP-s |
350.2kb | Sense | Chr. 8 - 77250364 - 77517953 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138650.1, AK162930.1 transcripts.
Set Membership: |
|

Gm10649
Genomic Map