Assay Details
| Gene Symbol: Stmn1-rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029430.1 | 1 | 450 | 86 | ||
| GenBank mRNA | CF896624.1 | - | 1 | 193 | 86 | |
| CF897466.1 | - | 1 | 163 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oxysterol binding protein-like 10 | Osbpl10 |
4933433D06Rik, C820004B04Rik, OPR-10 |
-47.8kb | Anti-sense | Chr. 9 - 114978583 - 115233273 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930428G15 gene | 4930428G15Rik | 54.1kb | Anti-sense | Chr. 9 - 115336161 - 115341924 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stmn1-rs1
Genomic Map