Assay Details
| Gene Symbol: Mir188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029574.1 | 1 | 28 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 362 | Mir362 |
Mirn362, mir-362, mmu-mir-362 |
-5.9kb | Sense | Chr. X - 7241982 - 7242046 on NCBI Build GRCm38 |
| Downstream | microRNA 532 | Mir532 |
Mirn532, mir-532, mmu-mir-532 |
346.0 | Sense | Chr. X - 7248402 - 7248497 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir188
Genomic Map