Assay Details
| Gene Symbol: Mir296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029646.1 | 1 | 40 | 51 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AL593857.10 | - | Chr. - - [-] on Build | 51 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10714 | Gm10714 | ENSMUSG00000074555 | -83.1kb | Sense | Chr. 2 - 174125038 - 174183918 on NCBI Build GRCm38 |
| Downstream | microRNA 298 | Mir298 |
Mirn298, mir-298, mmu-mir-298 |
379.0 | Sense | Chr. 2 - 174267504 - 174267585 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir296
Genomic Map