Assay Details
| Gene Symbol: Mirlet7c-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029729.1 | 1 | 48 | 81 | ||
| Gene Symbol: Lincppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110483.1 | 2 | 615 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | expressed sequence AU022754 | AU022754 | -112.9kb | Sense | Chr. 15 - 85581142 - 85593708 on NCBI Build GRCm38 | |
| Downstream | microRNA let7b | Mirlet7b |
Mirnlet7b, let-7b, mmu-let-7b |
622.0 | Sense | Chr. 15 - 85707319 - 85707403 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7c-2
Genomic Map