Assay Details
| Gene Symbol: Mir16-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029734.1 | 1 | 47 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
deleted in lymphocytic leukemia, 2 |
Dleu2 |
1810047A16Rik, 4932411A06Rik, 7730401J12Rik, AI197277, AI788824, Alt1, Gm9069, Leu2 |
Sense | Chr. 14 - 61602836 - 61682373 on NCBI Build GRCm38 | |
| Downstream | microRNA 15a | Mir15a |
Mirn15a, mir-15a, mmu-mir-15a |
55.0 | Sense | Chr. 14 - 61632027 - 61632110 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC1400002195.mm
This assay targets the same location as RTA transcript cluster(s) TC1500001845.rn
This assay targets the same location as RTA transcript cluster(s) TC1500001845.rn
Set Membership: |
|

Mir16-1
Genomic Map