Assay Details
| Gene Symbol: Mir93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029749.1 | 1 | 46 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 25 | Mir25 |
Mirn25, mir-25, mmu-mir-25 |
-119.0 | Sense | Chr. 5 - 138165321 - 138165404 on NCBI Build GRCm38 |
| Downstream | microRNA 106b | Mir106b |
Mirn106b, mir-106b |
127.0 | Sense | Chr. 5 - 138165737 - 138165818 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir93
Genomic Map