Assay Details
| Gene Symbol: Mir98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029753.1 | 1 | 56 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HECT, UBA and WWE domain containing 1 |
Huwe1 |
5430439H10Rik, AU041296, Arf-bp1, C430014N20Rik, C80292, Gm1718, Ib772, LASU1, Mule, Ureb1 |
Sense | Chr. X - 151800807 - 151935417 on NCBI Build GRCm38 | |
| Downstream | hydroxysteroid (17-beta) dehydrogenase 10 | Hsd17b10 |
17bHSD10, Ads9, ERAB, Hadh2 |
88.6kb | Sense | Chr. X - 152001896 - 152004442 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir98
Genomic Map