Assay Details
| Gene Symbol: Mir218-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029798.1 | 1 | 55 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | slit homolog 2 (Drosophila) | Slit2 |
Drad-1, E030015M03Rik, E130320P19Rik, Slil3, mKIAA4141, slit-2 |
Sense | Chr. 5 - 47983108 - 48306282 on NCBI Build GRCm38 | |
| Downstream | PARK2 co-regulated-like | Pacrgl | 4933428G09Rik | 148.3kb | Sense | Chr. 5 - 48372392 - 48388756 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir218-1
Genomic Map