Assay Details
| Gene Symbol: Mir7-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029826.1 | 1 | 38 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | de-etiolated homolog 1 (Arabidopsis) | Det1 | 2610034H20Rik | -40.9kb | Anti-sense | Chr. 7 - 78822959 - 78847382 on NCBI Build GRCm38 |
| Downstream | apoptosis enhancing nuclease | Aen |
2700083B06Rik, Isg20l1 |
7.6kb | Sense | Chr. 7 - 78895927 - 78908833 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7-2
Genomic Map