Assay Details
| Gene Symbol: Mir409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029913.1 | 1 | 39 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | miRNA containing gene | Mirg |
2810474H01Rik, AI852992, Gm2928, Meg9, mir-410 |
Sense | Chr. 12 - 109734981 - 109749458 on NCBI Build GRCm38 | |
| Downstream | microRNA 412 | Mir412 |
Mirn412, mir-412, mmu-mir-412 |
53.0 | Sense | Chr. 12 - 109743289 - 109743368 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir409
Genomic Map