Assay Details
| Gene Symbol: Mir367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030268.1 | 1 | 33 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 302d | Mir302d |
Mirn302d, mir-302d |
-44.0 | Sense | Chr. 3 - 127545624 - 127545689 on NCBI Build GRCm38 |
| Downstream |
zinc finger, GRF-type containing 1 |
Zgrf1 |
4930422G04Rik, AI448607 |
7.6kb | Sense | Chr. 3 - 127553436 - 127618023 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir367
Genomic Map