Assay Details
| Gene Symbol: Mir374b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030418.1 | 1 | 48 | 90 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AJ421479.1 | - | 16225 | Chr. - - [-] on Build | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 374c | Mir374c | mmu-mir-374c | Anti-sense | Chr. X - 103573085 - 103573133 on NCBI Build GRCm38 | |
| Downstream |
zinc finger, CCHC domain containing 13 |
Zcchc13 |
4930513O09Rik, Cnbp2 |
57.4kb | Anti-sense | Chr. X - 103630586 - 103631664 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir374b
Genomic Map