Assay Details
| Gene Symbol: Mir673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030438.1 | 1 | 52 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | maternally expressed 3 | Meg3 |
2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394 |
-261.0 | Sense | Chr. 12 - 109540996 - 109571729 on NCBI Build GRCm38 |
| Downstream | microRNA 493 | Mir493 |
Mirn493, mmu-mir-493 |
8.2kb | Sense | Chr. 12 - 109580233 - 109580315 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir673
Genomic Map