Assay Details
| Gene Symbol: Mir693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030466.1 | 1 | 43 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
polymerase (DNA directed), eta (RAD 30 related) |
Polh |
RAD30A, XPV |
-28.8kb | Anti-sense | Chr. 17 - 46171355 - 46202683 on NCBI Build GRCm38 |
| Downstream | polymerase (RNA) I polypeptide C | Polr1c |
40kDa, AA409007, AA959927, AL024089, Polr1e, RPA40, Rpo1-1 |
12.3kb | Anti-sense | Chr. 17 - 46243920 - 46248045 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mir693
Genomic Map