Assay Details
| Gene Symbol: Mir701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030482.1 | 1 | 54 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | checkpoint kinase 2 | Chek2 |
CHK2, Cds1, HUCDS1, Rad53 |
-130.0kb | Sense | Chr. 5 - 110839791 - 110874140 on NCBI Build GRCm38 |
| Downstream |
phosphatidylinositol transfer protein, beta |
Pitpnb | PI-TP-beta | 326.4kb | Sense | Chr. 5 - 111330697 - 111388361 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir701
Genomic Map