Assay Details
| Gene Symbol: Mir704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030485.1 | 1 | 38 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RNA, Y1 small cytoplasmic, Ro-associated |
Rny1 | -15.4kb | Sense | Chr. 6 - 47788070 - 47788180 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 786 | Zfp786 |
A730012O14Rik, Znf786 |
15.6kb | Sense | Chr. 6 - 47819263 - 47835920 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir704
Genomic Map