Assay Details
| Gene Symbol: Mir710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030491.1 | 1 | 52 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35521 |
Gm35521 | -97.2kb | Sense | Chr. 8 - 64408654 - 64417100 on NCBI Build GRCm38 | |
| Downstream | carboxypeptidase E | Cpe |
CPH, Cph-1, Cph1, R74677, fat |
78.1kb | Sense | Chr. 8 - 64592542 - 64693040 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir710
Genomic Map