Assay Details
| Gene Symbol: Mir505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030499.1 | 1 | 43 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mcf.2 transforming sequence | Mcf2 |
B230117G22Rik, Dbl, Mcf-2 |
-215.2kb | Sense | Chr. X - 60055956 - 60179187 on NCBI Build GRCm38 |
| Downstream | predicted gene 7073 | Gm7073 | 41.6kb | Sense | Chr. X - 60436052 - 60456195 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir505
Genomic Map