Assay Details
| Gene Symbol: Mir652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030523.1 | 1 | 45 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | acyl-CoA synthetase long-chain family member 4 | Acsl4 |
9430020A05Rik, ACS4, AU018108, Facl4, Lacs4 |
-348.2kb | Anti-sense | Chr. X - 142317993 - 142390798 on NCBI Build GRCm38 |
| Downstream |
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1 |
Ammecr1 | 6230420G18Rik | 114.4kb | Anti-sense | Chr. X - 142853474 - 142966728 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir652
Genomic Map