Assay Details
| Gene Symbol: E330033B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030690.1 | 1 | 949 | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4833422M21 gene | 4833422M21Rik | 4933406G12Rik | -13.9kb | Sense | Chr. 15 - 96231870 - 96254684 on NCBI Build GRCm38 |
| Downstream |
AT rich interactive domain 2 (ARID, RFX-like) |
Arid2 |
1700124K17Rik, 4432409D24Rik, zipzap/p200 |
12.2kb | Anti-sense | Chr. 15 - 96287522 - 96405463 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

E330033B04Rik
Genomic Map