Assay Details
| Gene Symbol: Plscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168497.1 | NP_001161969.1 | 3 - 4 | 553 | 70 | |
| NM_023564.4 | NP_076053.1 | 3 - 4 | 477 | 70 | ||
| NR_033133.1 | 2 - 3 | 666 | 70 | |||
| GenBank mRNA | AF159850.1 | - | 2 - 3 | 415 | 70 | |
| AK011716.1 | - | 3 - 4 | 475 | 70 | ||
| AK029154.1 | - | 2 - 3 | 633 | 70 | ||
| AK075841.1 | - | 3 - 4 | 554 | 70 | ||
| AK166288.1 | - | 3 - 4 | 460 | 70 | ||
| AK172635.1 | - | 3 - 4 | 454 | 70 | ||
| BC020143.1 | - | 2 - 3 | 412 | 3967872 | 70 | |
More Information
Set Membership: |
|

Plscr3
Genomic Map