Assay Details
| Gene Symbol: Gm17821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033146.1 | 1 | 10748 | 98 | ||
| GenBank mRNA | CJ164117.1 | - | 1 | 364 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAM domain containing glycosylphosphatidylinositol anchor 2 | Mdga2 |
6720489L24Rik, 9330209L04Rik, Adp, Mamdc1 |
-434.2kb | Sense | Chr. 12 - 66466059 - 67222549 on NCBI Build GRCm38 |
| Downstream | ribosomal protein S29 | Rps29 | 1.5mb | Sense | Chr. 12 - 69157722 - 69159186 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm17821
Genomic Map