Assay Details
| Gene Symbol: 1700069L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033216.1 | 1 - 2 | 139 | 96 | ||
| GenBank mRNA | AK006923.1 | - | 1 - 2 | 116 | 96 | |
| AK007096.1 | - | 1 - 2 | 140 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chemokine-like receptor 1 | Cmklr1 |
ChemR23, DEZ, Gpcr27, mcmklr1 |
-73.3kb | Sense | Chr. 5 - 113612355 - 113650450 on NCBI Build GRCm38 |
| Downstream | FIC domain containing | Ficd |
D5Ertd40e, Hype |
10.9kb | Anti-sense | Chr. 5 - 113735677 - 113740607 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006923.1 transcripts.
Set Membership: |
|

1700069L16Rik
Genomic Map