Assay Details
| Gene Symbol: Gm5627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033301.1 | 1 - 2 | 287 | 99 | ||
| GenBank mRNA | AK041874.1 | - | 1 - 2 | 290 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | angiomotin-like 2 | Amotl2 |
AW549739, C79691, Lccp, MASCOT, mKIAA0989 |
-6.2kb | Sense | Chr. 9 - 102717294 - 102733418 on NCBI Build GRCm38 |
| Downstream | receptor-like tyrosine kinase | Ryk |
AW536699, ERK-3, Vik |
87.3kb | Sense | Chr. 9 - 102834920 - 102908307 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041874.1 transcripts.
Set Membership: |
|

Gm5627
Genomic Map