Assay Details
| Gene Symbol: Gm5627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033301.1 | 2 - 3 | 393 | 65 | ||
| GenBank mRNA | AK041874.1 | - | 2 - 3 | 396 | 65 | |
| BC147467.1 | - | 2 - 3 | 147 | 9056109 | 65 | |
| BC147470.1 | - | 2 - 3 | 147 | 9056112 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | angiomotin-like 2 | Amotl2 |
AW549739, C79691, Lccp, MASCOT, mKIAA0989 |
-14.1kb | Sense | Chr. 9 - 102717294 - 102733418 on NCBI Build GRCm38 |
| Downstream | receptor-like tyrosine kinase | Ryk |
AW536699, ERK-3, Vik |
85.4kb | Sense | Chr. 9 - 102834920 - 102908307 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5627
Genomic Map