Assay Details
| Gene Symbol: Taz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173547.2 | NP_001167018.1 | 4 - 5 | 656 | 67 | |
| NM_001242615.2 | NP_001229544.1 | 4 - 5 | 656 | 67 | ||
| NM_001242616.2 | NP_001229545.1 | 4 - 5 | 656 | 67 | ||
| NM_181516.6 | NP_852657.1 | 4 - 5 | 656 | 67 | ||
| GenBank mRNA | AK075615.1 | - | 4 - 5 | 613 | 67 | |
| AK076481.1 | - | 4 - 5 | 624 | 67 | ||
| AK160327.1 | - | 4 - 5 | 625 | 67 | ||
| AK170662.1 | - | 4 - 5 | 625 | 67 | ||
| AK188231.1 | - | 2 - 3 | 114 | 67 | ||
| AY231459.1 | - | 4 - 5 | 375 | 67 | ||
| AY231460.1 | - | 4 - 5 | 375 | 67 | ||
| AY258040.1 | - | 4 - 5 | 375 | 67 | ||
| AY258041.1 | - | 4 - 5 | 375 | 67 | ||
| BC015305.1 | - | 4 - 5 | 621 | 4221041 | 67 | |
More Information
Set Membership: |
|

Taz
Genomic Map