Assay Details
| Gene Symbol: Taz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242616.2 | NP_001229545.1 | 6 - 7 | 783 | 99 | |
| NM_181516.6 | NP_852657.1 | 6 - 7 | 783 | 99 | ||
| GenBank mRNA | AK075615.1 | - | 6 - 7 | 740 | 99 | |
| AK076481.1 | - | 6 - 7 | 751 | 99 | ||
| AK160327.1 | - | 6 - 7 | 752 | 99 | ||
| AK188231.1 | - | 4 - 5 | 241 | 99 | ||
| AY231459.1 | - | 6 - 7 | 502 | 99 | ||
| AY231460.1 | - | 6 - 7 | 502 | 99 | ||
| AY258040.1 | - | 6 - 7 | 502 | 99 | ||
| AY258041.1 | - | 6 - 7 | 502 | 99 | ||
| BC015305.1 | - | 6 - 7 | 748 | 4221041 | 99 | |
| EF524069.1 | - | 5 - 6 | 382 | 99 | ||
More Information
Set Membership: |
|

Taz
Genomic Map